Learn More
Medchemexpress LLC Dicobalt edetate | 36499-65-7 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HY12233010MG
Dicobalt edetate is a cobalt complex used as an antidote for cyanide poisoning and is supplied as a high-purity research reagent. It forms stable complexes with cyanide ions and is typically provided in small laboratory pack sizes.
- Used as a cyanide antidote and research reagent.
- Molecular formula C10H12Co2N2O8.
- Molecular weight 406.08 g/mol.
- High purity (99.9%).
- Typical pack size 10 mg for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.