Learn More
Selleck Chemical LLC Dicoumarol-50mg
SDP
Catalog No. 502389679
Encompass_Preferred
Dicoumarol is a competitive NADPH quinone oxidoreductase (NQO1) inhibitor,and used as an anticoagulant by interfering with the metabolism of vitamin K.
Catalog No. 50-238-9679 Supplier Selleck Chemical LLC Supplier No. S429950MG
Dicoumarol is a competitive NADPH quinone oxidoreductase (NQO1) inhibitor,and used as an anticoagulant by interfering with the metabolism of vitamin K.