Learn More
Selleck Chemical LLC Dicoumarol S4299-200mg
SDP
Catalog No. 503112182
Encompass_Preferred
Dicoumarol is a competitive NADPH quinone oxidoreductase (NQO1) inhibitor and used as an anticoagulant by interfering with the metabolism of vitamin K
Catalog No. 50-311-2182 Supplier Selleck Chemical LLC Supplier No. S4299200MG
Dicoumarol is a competitive NADPH quinone oxidoreductase (NQO1) inhibitor and used as an anticoagulant by interfering with the metabolism of vitamin K