Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dicyandiamide-13C2,15N2 | MFCD00008066 | 99.9% | 88.05 | 13C2H4N215N2 | 1mg
SDP

Catalog No. 5000269664
Encompass_Preferred

Dicyandiamide-13C2 15N2 is the 15N2 labeled Dicyandiamide

Catalog No. 50-002-69664 Supplier Medchemexpress LLC Supplier No. HYY0674S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dicyandiamide-13C2 15N2 is the 15N2 labeled Dicyandiamide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.