Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dicyandiamide

Catalog No. D766093KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D766093KG 3 kg
D766091KG 1 kg
D7660925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D766093KG Supplier Sigma Aldrich Supplier No. D766093KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dicyandiamide

Specifications

CAS 461-58-5
Melting Point 208°C to 211°C (lit.)
Molecular Formula C2H4N4
Linear Formula NH2C(=NH)NHCN
MDL Number MFCD00008066
Quantity 3 kg
Synonym DCD; Cyanoguanidine; Dicyanodiamide
Molecular Weight (g/mol) 84.08
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.