Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyandiamide, 99%

Catalog No. AAA1045136
Click to view available options
Quantity:
250 g
500 g
2500 g

Catalog No. AAA1045136

Supplier: Thermo Scientific Chemicals A1045136

Only null left
Add to Cart

CAS: 461-58-5 | C2H4N4 | 84.08 g/mol

Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

Solubility
Soluble in Water (32g/L ) at 20°C

Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
IUPAC Name 2-cyanoguanidine
SMILES NC(N)=NC#N

Specifications

Melting Point 208°C to 211°C
Density 1.4 g/mL
Assay Percent Range 99%
Quantity 500 g
Beilstein 605637
Merck Index 14,3092
Solubility Information Soluble in Water (32g/L ) at 20°C
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material Dicyandiamide
EINECSNumber 207-312-8
RTECSNumber ME9950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.