Learn More
Abcam 9-(2,2-Dicyanovinyl)julolidine, 5MG
Catalog No. 5051306
Encompass_Preferred
MW 249.31 Da, Purity >97%. Fluorescent rotor probe for staining proteins and cellular structures. Quantum yield increases with decreasing free rotation upon binding. Used to study kinetics of beta-amyloid fibril formation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-513-06 Supplier Abcam Supplier No. AB2748755MG
MW 249.31 Da, Purity >97%. Fluorescent rotor probe for staining proteins and cellular structures. Quantum yield increases with decreasing free rotation upon binding. Used to study kinetics of beta-amyloid fibril formation.
The product is subject to the following: Abcam Restricted Use Statement