Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexyl Ketone 97.0+%, TCI America™
SDP

Catalog No. D2258500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2258500ML 500 mL
D225825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2258500ML Supplier TCI America Supplier No. D2258500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119-60-8
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.318
MDL Number MFCD00040417
InChI Key TWXWPPKDQOWNSX-UHFFFAOYSA-N
Synonym Dodecahydrobenzophenone
PubChem CID 8403
IUPAC Name dicyclohexylmethanone
SMILES C1CCC(CC1)C(=O)C2CCCCC2

Specifications

Color Yellow
Boiling Point 135°C
Quantity 500 mL
Formula Weight 194.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclohexyl Ketone
RTECSNumber OB1804500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.