Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexylamine, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
2.5L
Packaging:
Glass bottle
101-83-7
C12H23N
181.32
MFCD00011658
XBPCUCUWBYBCDP-UHFFFAOYSA-N
dicyclohexylamine,dcha,dicha,cyclohexanamine, n-cyclohexyl,n,n-dicyclohexylamine,dodecahydrodiphenylamine,dicyklohexylamin,n,n-diclohexylamine,n-cyclohexyl-cyclohexylamine,dicyklohexylamin czech
7582
CHEBI:34694
N-cyclohexylcyclohexanamine
C1CCC(CC1)NC2CCCCC2
101-83-7
C12H23N
181.32
MFCD00011658
XBPCUCUWBYBCDP-UHFFFAOYSA-N
dicyclohexylamine,dcha,dicha,cyclohexanamine, n-cyclohexyl,n,n-dicyclohexylamine,dodecahydrodiphenylamine,dicyklohexylamin,n,n-diclohexylamine,n-cyclohexyl-cyclohexylamine,dicyklohexylamin czech
7582
CHEBI:34694
N-cyclohexylcyclohexanamine
C1CCC(CC1)NC2CCCCC2
Specific Gravity 0.91
Viscosity 7.4 mPa.s (20°C)
Refractive Index 1.4832 to 1.4852
Water 0.3% max.
Infrared Spectrum Authentic
Flash Point 103°C
Packaging Glass bottle
Quantity 500mL
Melting Point -2.0°C
Color White
Density 0.9100g/mL
Boiling Point 256.0°C
Assay Percent Range 99% min. (GC)
Linear Formula (C6H11)2NH
Beilstein 12,6
Fieser 01,231
Merck Index 15,3106
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 181.32
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Dicyclohexylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 202-980-7