Learn More
Medchemexpress LLC Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid | 33164-70-4 | 416.62 | C21H40N2O4S | 10 MG
SDP
Catalog No. 5000285111
Encompass_Preferred
Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid is the dicyclohexylammonium salt of a crotonaldehyde metabolite supplied as a reference standard for analytical and research applications. It is intended for metabolite identification, method development, and quantitative assay support. Lot-specific purity and recommended storage conditions are provided on the Certificate of Analysis.
Catalog No. 50-002-85111 Supplier Medchemexpress LLC Supplier No. HYW58774210MG
Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid is the dicyclohexylammonium salt of a crotonaldehyde metabolite supplied as a reference standard for analytical and research applications. It is intended for metabolite identification, method development, and quantitative assay support. Lot-specific purity and recommended storage conditions are provided on the Certificate of Analysis.