Learn More
Medchemexpress LLC Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid | 33164-70-4 | >95.0% | 416.62 g/mol | C21H40N2O4S | 1 MG
SDP
Catalog No. 5000283969
Encompass_Preferred
Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid is a drug metabolite provided as an analytical reference standard for laboratory and analytical research. It is intended for method development, chromatographic assay validation, and metabolite identification workflows.
  • Provided as a high-purity analytical standard (>95% by HPLC).
  • Suitable for method development and metabolite identification.
  • Supplied in small milligram sizes for trace-level analysis.
  • Molecular weight 416.62 g/mol and formula C21H40N2O4S for reference.
  • Certificate of analysis available with storage and assay details.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-83969 Supplier Medchemexpress LLC Supplier No. HYW5877421MG
Dicyclohexylammonium 3-hydroxy-1-methylpropylmercapturic acid is a drug metabolite provided as an analytical reference standard for laboratory and analytical research. It is intended for method development, chromatographic assay validation, and metabolite identification workflows.
  • Provided as a high-purity analytical standard (>95% by HPLC).
  • Suitable for method development and metabolite identification.
  • Supplied in small milligram sizes for trace-level analysis.
  • Molecular weight 416.62 g/mol and formula C21H40N2O4S for reference.
  • Certificate of analysis available with storage and assay details.
  • Intended for research use only; not for human or clinical use.