Learn More
Strem, An Ascensus Company CAS# 16523-54-9. 5G. Dicyclohexylchlorophosphine, min. 98%. MFCD00014294
SDP
Supplier:  Strem, An Ascensus Company 1510505G
Encompass
CAS# 16523-54-9. 5G. Dicyclohexylchlorophosphine, min. 98%. MFCD00014294. Molecular Formula: (C6H11)2PCl. Molecular Weight: 232.74. Color/Form: colorless to pale yellow, cloudy liq. Stability: air sensitive, moisture sensitive. Strem# 15-1050. www.strem.com/catalog/v3/15-1050/
Catalog No. 50-196-3589