Learn More
Apexbio Technology LLC Dicyclomine HCl 67-92-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096236
Encompass_Preferred
Dicyclomine Hydrochloride (CAS 67-92-5) is an orally active muscarinic acetylcholine receptor antagonist that primarily interacts with the M1 and M2 receptor subtypes It exhibits high binding affinity for the M1 subtype (Ki 5 nM) and moderate affinity for M2 receptors (Ki 55 nM) By inhibiting these receptors Dicyclomine HCl is utilized in studies investigating muscarinic receptor-mediated signal transduction Additionally it is commonly employed in pharmacological and pathophysiological research involving gastrointestinal smooth muscle spasm and related mechanisms
Catalog No. 50-000-96236 Supplier Apexbio Technology LLC Supplier No. A84095.1
Dicyclomine Hydrochloride (CAS 67-92-5) is an orally active muscarinic acetylcholine receptor antagonist that primarily interacts with the M1 and M2 receptor subtypes It exhibits high binding affinity for the M1 subtype (Ki 5 nM) and moderate affinity for M2 receptors (Ki 55 nM) By inhibiting these receptors Dicyclomine HCl is utilized in studies investigating muscarinic receptor-mediated signal transduction Additionally it is commonly employed in pharmacological and pathophysiological research involving gastrointestinal smooth muscle spasm and related mechanisms