Learn More
Medchemexpress LLC Dicyclomine hydrochloride | 67-92-5 | 98.1% | 100 MG
SDP
Catalog No. 5000412066
Encompass_Preferred
Dicyclomine hydrochloride is a potent, orally active antagonist of muscarinic cholinergic receptors, showing high affinity for M1 (Ki=5.1 nM) and M2 (Ki=54.6 nM) subtypes. It acts as an antispasmodic agent, relieving smooth muscle spasms in the gastrointestinal tract in vivo.
  • An orally active antagonist of muscarinic cholinergic receptors.
  • Demonstrates high affinity for both M1 and M2 muscarinic receptor subtypes (Ki values: M1=5.1 nM, M2=54.6 nM).
  • Functions as an antispasmodic agent.
  • Relieves smooth muscle spasms in the gastrointestinal tract in vivo.
  • May exacerbate cognitive impairments and lead to behavioral impairments in spatial tasks in animal models.
Catalog No. 50-004-12066 Supplier Medchemexpress LLC Supplier No. HYB1339100MG
Dicyclomine hydrochloride is a potent, orally active antagonist of muscarinic cholinergic receptors, showing high affinity for M1 (Ki=5.1 nM) and M2 (Ki=54.6 nM) subtypes. It acts as an antispasmodic agent, relieving smooth muscle spasms in the gastrointestinal tract in vivo.
  • An orally active antagonist of muscarinic cholinergic receptors.
  • Demonstrates high affinity for both M1 and M2 muscarinic receptor subtypes (Ki values: M1=5.1 nM, M2=54.6 nM).
  • Functions as an antispasmodic agent.
  • Relieves smooth muscle spasms in the gastrointestinal tract in vivo.
  • May exacerbate cognitive impairments and lead to behavioral impairments in spatial tasks in animal models.