Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dicyclomine (hydrochloride) | 67-92-5 | 98.1% | 500 MG
SDP

Catalog No. 5000413512
Encompass_Preferred

Dicyclomine hydrochloride is a potent and orally active antagonist of muscarinic cholinergic receptors. It exhibits high affinity for both muscarinic M1 receptor subtype (Ki=5.1 nM) and M2 receptor subtype (Ki=54.6 nM) in brush-border membrane and basal plasma membranes, respectively. This compound functions as an antispasmodic agent, relieving smooth muscle spasm of the gastrointestinal tract in vivo. It is intended for research use only and not for sale to patients.

  • Potent muscarinic cholinergic receptors antagonist.
  • High affinity for muscarinic M1 (Ki=5.1 nM) and M2 (Ki=54.6 nM) receptor subtypes.
  • Acts as an antispasmodic agent that relieves smooth muscle spasm of the gastrointestinal tract in vivo.
  • Orally active.
  • Purity: 98.09%.
  • Appearance: Solid, white to off-white.
  • Soluble in H2O (50 mg/mL) and DMSO (33.33 mg/mL).

Catalog No. 50-004-13512 Supplier Medchemexpress LLC Supplier No. HYB1339500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dicyclomine hydrochloride is a potent and orally active antagonist of muscarinic cholinergic receptors. It exhibits high affinity for both muscarinic M1 receptor subtype (Ki=5.1 nM) and M2 receptor subtype (Ki=54.6 nM) in brush-border membrane and basal plasma membranes, respectively. This compound functions as an antispasmodic agent, relieving smooth muscle spasm of the gastrointestinal tract in vivo. It is intended for research use only and not for sale to patients.

  • Potent muscarinic cholinergic receptors antagonist.
  • High affinity for muscarinic M1 (Ki=5.1 nM) and M2 (Ki=54.6 nM) receptor subtypes.
  • Acts as an antispasmodic agent that relieves smooth muscle spasm of the gastrointestinal tract in vivo.
  • Orally active.
  • Purity: 98.09%.
  • Appearance: Solid, white to off-white.
  • Soluble in H2O (50 mg/mL) and DMSO (33.33 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.