Learn More
Cayman Chemical Dicyclomine (hydrochloride), Purity: ≥98%, CAS: 67-92-5, 500 mg
SDP
Catalog No. 502964468
Encompass_Preferred
A M1 and M2 muscarinic acetylcholine receptor antagonist (Kis = 3.16 and 44.7 nM, respectively); inhibits carbachol-stimulated inositol phosphate accumulation in guinea pig cortex (Ki = 12 nM; pA2 = 7.88); inhibits potassium-induced contraction of ileum, colon, and duodenum in anesthetized rats by 21.85, 30.81, and 37.86%, respectively (10 mg/kg, i.v.)
Catalog No. 50-296-4468 Supplier Cayman Chemical Supplier No. 23867500 MG
A M1 and M2 muscarinic acetylcholine receptor antagonist (Kis = 3.16 and 44.7 nM, respectively); inhibits carbachol-stimulated inositol phosphate accumulation in guinea pig cortex (Ki = 12 nM; pA2 = 7.88); inhibits potassium-induced contraction of ileum, colon, and duodenum in anesthetized rats by 21.85, 30.81, and 37.86%, respectively (10 mg/kg, i.v.)