Learn More
TARGETMOL CHEMICALS INC DiD perchlorate 10MG
SDP
Catalog No. 502879698
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DiD perchlorate is a lipophilic cyanine dye that can be used in cells tracking. Purity ≥98%
Catalog No. 50-287-9698 Supplier TARGETMOL CHEMICALS INC Supplier No. T1511810MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DiD perchlorate is a lipophilic cyanine dye that can be used in cells tracking. Purity ≥98%