Learn More
Medchemexpress LLC HY-D1028 5mg Medchemexpress, DiD perchlorate CAS:127274-91-3 Purity:>98%
SDP
Catalog No. 501970195
Encompass_Preferred
Medchemexpress, HY-D1028 5mg DiD perchlorate CAS:127274-91-3 DiD perchlorate is a lipophilic cyanine dye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0195 Supplier Medchemexpress LLC Supplier No. HYD10285MG
Medchemexpress, HY-D1028 5mg DiD perchlorate CAS:127274-91-3 DiD perchlorate is a lipophilic cyanine dye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.