Learn More
Selleck Chemical LLC Didanosine 50mg 69655-05-6
SDP
Catalog No. 505550
Encompass_Preferred
Didanosine is a reverse transcriptase inhibitor with an IC50 of 0.49 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-555-0 Supplier Selleck Chemical LLC Supplier No. S170250MG
Didanosine is a reverse transcriptase inhibitor with an IC50 of 0.49 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.