Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didecyl phthalate, MilliporeSigma™ Supelco™

Catalog No. 111015678 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5678 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5678 Supplier MilliporeSigma Supelco Supplier No. 36933250MG
Add to Cart
Add to Cart

Didecyl phthalate belongs to the class of phthalate esters widely used as plasticizers in plastic films.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 84-77-5
Flash Point 200°C - Closed Cup
Physical Form Neat
Linear Formula C6H4 [COO(CH2)9 CH3]2
MDL Number MFCD00041915
Type Analytical Standard
Quantity 250 mg
Formula Weight 446.66
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.