Learn More
Medchemexpress LLC Didecyldimethylammonium bromide | 2390-68-3 | MFCD00012194 | 98.0% | 406.53 | C22H48BrN | 250 MG
SDP
Catalog No. 5000282636
Encompass_Preferred
Didecyldimethylammonium bromide is a quaternary ammonium compound used as a biochemical reagent and broad-spectrum biocide for laboratory research. It is supplied as a high-purity solid suitable for formulation, analytical, and microbiological applications; handle according to standard research chemical safety guidance.
  • Quaternary ammonium biocide effective against bacteria and fungi.
  • High purity: 98.0%.
  • Available in small research quantities, including 250 mg.
  • Molecular weight 406.53 and formula C22H48BrN for formulation calculations.
  • Intended for research use only; follow safety data sheet precautions.
Catalog No. 50-002-82636 Supplier Medchemexpress LLC Supplier No. HYW127652250MG
Didecyldimethylammonium bromide is a quaternary ammonium compound used as a biochemical reagent and broad-spectrum biocide for laboratory research. It is supplied as a high-purity solid suitable for formulation, analytical, and microbiological applications; handle according to standard research chemical safety guidance.
  • Quaternary ammonium biocide effective against bacteria and fungi.
  • High purity: 98.0%.
  • Available in small research quantities, including 250 mg.
  • Molecular weight 406.53 and formula C22H48BrN for formulation calculations.
  • Intended for research use only; follow safety data sheet precautions.