Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Didecyldimethylammonium chloride | 7173-51-5 | MFCD00066262 | 98.0% | 362.1 g/mol | C22H48ClN | 1 G
SDP

Catalog No. 5000208826
Encompass_Preferred

Didecyldimethylammonium chloride is a dialkyl quaternary ammonium chloride used as a biocidal agent with broad-spectrum bactericidal, virucidal, and fungicidal activity. It is supplied as a high-purity reagent for laboratory and formulation applications.

  • Quaternary ammonium compound with broad-spectrum biocidal activity.
  • CAS number 7173-51-5.
  • Molecular formula C22H48ClN and molecular weight 362.1 g/mol.
  • Purity typically 98.0% for laboratory-grade material.
  • Used in disinfectant formulation research and antimicrobial testing.
  • Provided in small quantities suitable for research and development.

Catalog No. 50-002-08826 Supplier Medchemexpress LLC Supplier No. HYW0421811G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Didecyldimethylammonium chloride is a dialkyl quaternary ammonium chloride used as a biocidal agent with broad-spectrum bactericidal, virucidal, and fungicidal activity. It is supplied as a high-purity reagent for laboratory and formulation applications.

  • Quaternary ammonium compound with broad-spectrum biocidal activity.
  • CAS number 7173-51-5.
  • Molecular formula C22H48ClN and molecular weight 362.1 g/mol.
  • Purity typically 98.0% for laboratory-grade material.
  • Used in disinfectant formulation research and antimicrobial testing.
  • Provided in small quantities suitable for research and development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.