Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didecyldimethylammonium chloride, MilliporeSigma™ Supelco™

Catalog No. 111018919 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8919 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8919 Supplier MilliporeSigma Supelco Supplier No. 34466250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7173-51-5
Flash Point 26.5°C - Closed Cup
Linear Formula C22H48 ClN
MDL Number MFCD00066262
Synonym N-Decyl-N,N-dimethyl-1-decanaminium chloride; DDAC
Type Analytical Standard
Quantity 250 mg
Formula Weight 362.08
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.