Learn More
Medchemexpress LLC Didesmethyl cariprazine | 839712-25-3 | MFCD34469127 | 99.6% | 399.36 | C19H28Cl2N4O | 5 MG
SDP
Catalog No. 5000213802
Encompass_Preferred
Didesmethyl cariprazine is the predominant circulating, orally active metabolite of cariprazine used in pharmacology research to probe dopaminergic and serotonergic mechanisms. It crosses the blood-brain barrier and modulates dopamine and serotonin receptors, making it useful for receptor profiling, metabolism studies, and in vivo pharmacokinetic investigations.
  • Orally active, blood-brain barrier permeable compound.
  • Partial agonist at D2 and D3 receptors.
  • Full agonist at 5-HT1A and antagonist at 5-HT2B.
  • Suitable for in vitro receptor pharmacology studies.
  • Applicable to in vivo pharmacokinetic and metabolism research.
  • Provided as a solid with high reported purity for research use.
Catalog No. 50-002-13802 Supplier Medchemexpress LLC Supplier No. HY1006585MG
Didesmethyl cariprazine is the predominant circulating, orally active metabolite of cariprazine used in pharmacology research to probe dopaminergic and serotonergic mechanisms. It crosses the blood-brain barrier and modulates dopamine and serotonin receptors, making it useful for receptor profiling, metabolism studies, and in vivo pharmacokinetic investigations.
  • Orally active, blood-brain barrier permeable compound.
  • Partial agonist at D2 and D3 receptors.
  • Full agonist at 5-HT1A and antagonist at 5-HT2B.
  • Suitable for in vitro receptor pharmacology studies.
  • Applicable to in vivo pharmacokinetic and metabolism research.
  • Provided as a solid with high reported purity for research use.