Learn More
Cayman Chemical Didanosine-d2, Purity: ≥99% deuterated forms (d1-d2), 1 mg
SDP
Catalog No. 502992445
Encompass_Preferred
An internal standard for the quantification of didanosine by GC- or LC-MS
Catalog No. 50-299-2445 Supplier Cayman Chemical Supplier No. 300781 MG
An internal standard for the quantification of didanosine by GC- or LC-MS