Learn More
Cayman Chemical DIdnosIn-d2 500ug
SDP
Catalog No. 502988348
Encompass_Preferred
An internal standard for the quantification of didanosine by GC- or LC-MS
Catalog No. 50-298-8348 Supplier Cayman Chemical Supplier No. 30078500 UG
An internal standard for the quantification of didanosine by GC- or LC-MS