Learn More
Aviva Systems Biology Corp DIDO1 Antibody - C-terminal region (ARP39544_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39544_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DIDO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DIDO1. Alternate names: BYE1, C20orf158, DATF1, DIDO2, DIDO3, DIO-1, DIO1, DKFZp434P1115, FLJ11265, KIAA0333, MGC16140, dJ885L7.8, DATF-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57079