Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Didodecyl 3,3'-thiodipropionate

Catalog No. D128406250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D128406250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D128406250G Supplier Sigma Aldrich Supplier No. D128406250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Didodecyl 3,3'-thiodipropionate

Specifications

CAS 123-28-4
Melting Point 40°C to 42°C
Density 0.915 g/mL (at 25°C)
Molecular Formula C30H58O4S
Linear Formula S[CH2CH2CO2(CH2)11 CH3]2
Quantity 250 g
MDL Number MFCD00026589
Synonym Thiodipropionic acid dilauryl ester
Molecular Weight (g/mol) 514.84
Percent Purity >97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.