Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didodecyl Succinate 93.0+%, TCI America™
SDP

Catalog No. S025525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S025525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S025525G Supplier TCI America Supplier No. S025525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5980-15-4
Molecular Formula C28H54O4
Molecular Weight (g/mol) 454.74
MDL Number MFCD00059284
InChI Key JBJMZCVEBLDYCA-UHFFFAOYSA-N
Synonym Succinic Acid Dilauryl Ester, Dilauryl Succinate, Succinic Acid Didodecyl Ester
PubChem CID 80082
IUPAC Name 1,4-didodecyl butanedioate
SMILES CCCCCCCCCCCCOC(=O)CCC(=O)OCCCCCCCCCCCC

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 25 g
Formula Weight 454.74
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Didodecyl Succinate
EINECSNumber (2)-0901
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.