Learn More
Medchemexpress LLC Didodecylamine (N-dodecyldodecan-1-amine) | 3007-31-6 | MFCD00041918 | >=97.0% | 353.67 g/mol | C24H51N | 50 G
SDP
Supplier:  Medchemexpress LLC HYW14252550G
Encompass_Preferred
Didodecylamine is a secondary dialkylamine (N-dodecyldodecan-1-amine) used as a reagent and surfactant-type intermediate in organic synthesis, formulation development, and surface-modification research. It contains two saturated C12 alkyl chains and is typically handled as a hydrophobic, low-polarity amine for laboratory applications.
  • Secondary dialkylamine with two C12 chains.
  • Hydrophobic, low-polarity organic amine.
  • Suitable for surfactant, phase-transfer, and coupling applications.
  • Available in laboratory pack sizes for research use.
  • Typical supplier purity ≥97% for research-grade material.
Catalog No. 50-002-83542