Learn More
Medchemexpress LLC Didodecylamine (N-dodecyldodecan-1-amine) | 3007-31-6 | MFCD00041918 | 98.0% | 353.67 g/mol | C24H51N | 5 G
SDP
Catalog No. 5000283780
Encompass_Preferred
Didodecylamine (N-dodecyldodecan-1-amine, CAS 3007-31-6) is a long-chain secondary alkylamine used as a surfactant, intermediate, and reagent in organic synthesis and materials research. Supplied at approximately 98% purity, with molecular formula C24H51N and molecular weight 353.67 g/mol, it is intended for laboratory research applications and bench-scale use.
  • High purity suitable for research-grade applications.
  • Long hydrophobic C12-C12 chain provides surfactant and phase-transfer properties.
  • Useful as an intermediate in organic synthesis and materials functionalization.
  • Available in small research pack sizes for convenient bench use.
  • Handle according to standard laboratory safety and storage practices.
Catalog No. 50-002-83780 Supplier Medchemexpress LLC Supplier No. HYW1425255G
Didodecylamine (N-dodecyldodecan-1-amine, CAS 3007-31-6) is a long-chain secondary alkylamine used as a surfactant, intermediate, and reagent in organic synthesis and materials research. Supplied at approximately 98% purity, with molecular formula C24H51N and molecular weight 353.67 g/mol, it is intended for laboratory research applications and bench-scale use.
  • High purity suitable for research-grade applications.
  • Long hydrophobic C12-C12 chain provides surfactant and phase-transfer properties.
  • Useful as an intermediate in organic synthesis and materials functionalization.
  • Available in small research pack sizes for convenient bench use.
  • Handle according to standard laboratory safety and storage practices.