Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didodecyldimethylammonium bromide, 99%

Catalog No. AC407121000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407121000 100 g Plastic bottle
AC407120250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407121000 Supplier Thermo Scientific Chemicals Supplier No. 407121000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3282-73-3 | C26H56BrN | 462.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3282-73-3
Molecular Formula C26H56BrN
Molecular Weight (g/mol) 462.64
MDL Number MFCD00041969
InChI Key XRWMGCFJVKDVMD-UHFFFAOYSA-M
Synonym didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
PubChem CID 18669
IUPAC Name didodecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]

Specifications

Melting Point 161.0°C to 164.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3(CH2)11)2N(Br)(CH3)2
Quantity 100 g
Formula Weight 462.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Didodecyldimethylammonium bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 221-923-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.