Learn More
Cayman Chemical DIdodIum salt 50mg
SDP
Catalog No. 502970319
Encompass_Preferred
A stilbene sulfonate that inhibits anionic transport
Catalog No. 50-297-0319 Supplier Cayman Chemical Supplier No. 1612550 MG
A stilbene sulfonate that inhibits anionic transport