Learn More
Medchemexpress LLC HY-D0086 50mg Medchemexpress, DIDS sodium salt CAS:67483-13-0 Purity:>98%
SDP
Catalog No. 501933148
Encompass_Preferred
Medchemexpress, HY-D0086 50mg DIDS sodium salt CAS:67483-13-0 DIDS sodium salt is a dual ABCA1 and VDAC1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3148 Supplier Medchemexpress LLC Supplier No. HYD008650MG
Medchemexpress, HY-D0086 50mg DIDS sodium salt CAS:67483-13-0 DIDS sodium salt is a dual ABCA1 and VDAC1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.