Learn More
Medchemexpress LLC DIDS sodium salt | 67483-13-0 | 99.1% | 100 MG
SDP
Catalog No. 5000410037
Encompass_Preferred
DIDS sodium salt is a dual inhibitor of ABCA1 and VDAC1. It also inhibits RAD51, specifically targeting RAD51-mediated homologous pairing and strand exchange reactions. This compound inhibits anion exchange and binding to red blood cell membranes, and can suppress the activation of caspase-3 and -9, making it relevant for cancer research.
  • Is a dual inhibitor of ABCA1 and VDAC1.
  • Inhibits RAD51.
  • Inhibits RAD51-mediated homologous pairing and strand exchange reactions.
  • Inhibits anion exchange and binding to red blood cell membranes.
  • Inhibits the activation of caspase-3 and -9.
  • Can be used in cancer research.
Catalog No. 50-004-10037 Supplier Medchemexpress LLC Supplier No. HYD0086100MG
DIDS sodium salt is a dual inhibitor of ABCA1 and VDAC1. It also inhibits RAD51, specifically targeting RAD51-mediated homologous pairing and strand exchange reactions. This compound inhibits anion exchange and binding to red blood cell membranes, and can suppress the activation of caspase-3 and -9, making it relevant for cancer research.
  • Is a dual inhibitor of ABCA1 and VDAC1.
  • Inhibits RAD51.
  • Inhibits RAD51-mediated homologous pairing and strand exchange reactions.
  • Inhibits anion exchange and binding to red blood cell membranes.
  • Inhibits the activation of caspase-3 and -9.
  • Can be used in cancer research.