Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DIDS sodium salt | 67483-13-0 | MDL101114ZA | 99.1% | 498.48 | C16H8N2Na2O6S4 | 10 MG
SDP

Catalog No. 5000297322
Encompass_Preferred

DIDS sodium salt is the disodium form of 4,4'-diisothiocyanostilbene-2,2'-disulfonic acid, used as an anion-exchange inhibitor and modulator of VDAC and RAD51 in biochemical and cell biology research. It is supplied as a solid reagent with high solubility in DMSO for preparation of concentrated stock solutions.

  • High solubility in DMSO (125 mg/mL) for stock solution preparation.
  • Molecular weight 498.48 g/mol.
  • CAS number 67483-13-0.
  • Intended for research use; not for human or clinical use.
  • Recommended storage sealed at -20°C; in solution, -80°C for up to 6 months.

Catalog No. 50-002-97322 Supplier Medchemexpress LLC Supplier No. HYD008610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DIDS sodium salt is the disodium form of 4,4'-diisothiocyanostilbene-2,2'-disulfonic acid, used as an anion-exchange inhibitor and modulator of VDAC and RAD51 in biochemical and cell biology research. It is supplied as a solid reagent with high solubility in DMSO for preparation of concentrated stock solutions.

  • High solubility in DMSO (125 mg/mL) for stock solution preparation.
  • Molecular weight 498.48 g/mol.
  • CAS number 67483-13-0.
  • Intended for research use; not for human or clinical use.
  • Recommended storage sealed at -20°C; in solution, -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.