Learn More
Medchemexpress LLC HY-N2068 5mg Medchemexpress, Didymin CAS:14259-47-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20685MG
Encompass_Preferred
Medchemexpress, HY-N2068 5mg Didymin CAS:14259-47-3 Didymin, a dietary flavonoid glycoside from citrus fruits, possesses antioxidant properties. Didymin induces apoptosis by inhibiting N-Myc and upregulating RKIP in neuroblastoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0362