Learn More
Sklar Instruments Clamp, Dieffenbach Bulldog Clamp, Sklar 1-1/2 in., Blunt, Curved, Finely Serrated, Stainless Steel, EA.
SDP
Catalog No. 501182725 Shop All Sklar Instruments Products
Encompass_Preferred
Clamp, Dieffenbach Bulldog Clamp, Sklar, 1-1/2 in., Blunt, Curved, Finely Serrated, Stainless Steel, EA. Sklar's Diffenbach Bulldog Clamp are small, cross-action forceps most commonly used for clamping small vessels. They are often used in cardiovascular procedures. They have finely serrated jaws with rounded tips, that may be slightly curved or straight. A bulldog clamp may also be known as a serrefine. This product has curved jaws with an overall length of 1-1/2 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-118-2725 Supplier Sklar Instruments Supplier No. 173915
Clamp, Dieffenbach Bulldog Clamp, Sklar, 1-1/2 in., Blunt, Curved, Finely Serrated, Stainless Steel, EA. Sklar's Diffenbach Bulldog Clamp are small, cross-action forceps most commonly used for clamping small vessels. They are often used in cardiovascular procedures. They have finely serrated jaws with rounded tips, that may be slightly curved or straight. A bulldog clamp may also be known as a serrefine. This product has curved jaws with an overall length of 1-1/2 inches. Lifetime warranty against all manufacturer defects.