Learn More
Stoelting™ Dieffenbach Serrefine
SDP
Catalog No. 10000456 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Straight
Length (English):
1.2 in.
1.25 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Length (English)
10-000-456 Straight 1.2 in.
10-001-263 Curved 1.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-456 Supplier Stoelting™ Supplier No. 5210677P
Add to Cart
Add to Cart
Stoelting™ Dieffenbach Serrefine is a useful instrument for stopping blood flow to a particular organ of interest.
250 Grams of pressure
Specifications
For Use With (Application) Neuroscience
Length (English) 1.2 in.
Model Straight