Learn More
Perkin Elmer US LLC Dielectric Capillary for Spare Probe Assembly
Catalog No. 502113334
Encompass_Preferred
The innovative separation probes and holders facilitate chromatography directly at the ionization source. This minimizes dead volume effects in the system for superior signal-to-noise ratios, enhanced sensitivity and narrower peaks. Compatible with LC/MS systems using Flexar SQ300 or AxION 2 TOF MS.
Catalog No. 50-211-3334 Supplier Perkin Elmer US LLC Supplier No. MZ10505760180
The innovative separation probes and holders facilitate chromatography directly at the ionization source. This minimizes dead volume effects in the system for superior signal-to-noise ratios, enhanced sensitivity and narrower peaks. Compatible with LC/MS systems using Flexar SQ300 or AxION 2 TOF MS.