Learn More
Medchemexpress LLC Dienestrol-d2 | 1346606-45-8 | 500 UG
SDP
Catalog No. 5000411884
Encompass_Preferred
Dienestrol-d2 is a deuterium labeled Dienestrol. Stable heavy isotopes have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Used as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11884 Supplier Medchemexpress LLC Supplier No. HYB1403S500UG
Dienestrol-d2 is a deuterium labeled Dienestrol. Stable heavy isotopes have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Used as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs