Learn More
Medchemexpress LLC Dienogest-d6 | 65928-58-7 | 99.9% | C20H19D6NO2 | 1 MG
SDP
Catalog No. 5000432568
Encompass_Preferred
Dienogest-d6 (Synonyms: STS 557-d6) is a deuterium-labeled derivative of Dienogest (HY-B0084). Dienogest is known as an orally active progesterone receptor agonist that has applications in the study of endometriosis. This compound is intended for research use only and is not sold to patients. Dienogest-d6 has a molecular weight of 317.45 and appears as a white to off-white solid.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32568 Supplier Medchemexpress LLC Supplier No. HYB0084S21MG
Dienogest-d6 (Synonyms: STS 557-d6) is a deuterium-labeled derivative of Dienogest (HY-B0084). Dienogest is known as an orally active progesterone receptor agonist that has applications in the study of endometriosis. This compound is intended for research use only and is not sold to patients. Dienogest-d6 has a molecular weight of 317.45 and appears as a white to off-white solid.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.