Learn More
Alpco Ltd Dientamoeba fragilis PCR Kit
SDP
Catalog No. NC2284599
Encompass
The Dientamoeba fragilis multiplex real-time PCR assay is intended for the direct, qualitative detection of Dientamoeba fragilis in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284599 Supplier Alpco Ltd Supplier No. 86DFGHU100
The Dientamoeba fragilis multiplex real-time PCR assay is intended for the direct, qualitative detection of Dientamoeba fragilis in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.