Learn More
MSC CAM Spray Hot Water Pressure Washer, 4 GPM, 4000 PSI, Gas Engine, Diesel Fired
Catalog No. 502838423
Encompass
This portable hot water pressure washer delivers 4 GPM at 4000 PSI, designed for mobile use across various work locations. It features an industrial gas engine, a triplex plunger pump, and a diesel-fired burner system for hot water cleaning up to 195° F.
  • Mobile design with heavy duty frame, hose rack, and wand holder
  • 13 in wheels for navigating various terrains
  • Industrial gas engine and triplex plunger pump with ceramic plungers
  • Low pressure chemical injection
  • Unloader valve and secondary pressure pop off protect the system
  • Burner system with rust free fuel tank, Schedule 80 coil, and adjustable thermostat up to 195° F
  • Heavy duty trigger gun with dual wand for high pressure and detergent application
  • Quick change 0°, 15°, 25°, and 40° nozzles included
Catalog No. 50-283-8423 Supplier MSC Supplier No. 63821391
Add to Cart
Add to Cart
Request a Quote
This portable hot water pressure washer delivers 4 GPM at 4000 PSI, designed for mobile use across various work locations. It features an industrial gas engine, a triplex plunger pump, and a diesel-fired burner system for hot water cleaning up to 195° F.
  • Mobile design with heavy duty frame, hose rack, and wand holder
  • 13 in wheels for navigating various terrains
  • Industrial gas engine and triplex plunger pump with ceramic plungers
  • Low pressure chemical injection
  • Unloader valve and secondary pressure pop off protect the system
  • Burner system with rust free fuel tank, Schedule 80 coil, and adjustable thermostat up to 195° F
  • Heavy duty trigger gun with dual wand for high pressure and detergent application
  • Quick change 0°, 15°, 25°, and 40° nozzles included