Learn More
MSC CAM Spray Pressure Washer, Diesel Fired, Electric, 4 GPM, 2000 psi
Catalog No. 502838308
Encompass
This portable hot water pressure washer offers economical solutions for various applications. It features a frame with a hose and wand holder and rolls easily on two 13 in wheels.
  • Totally enclosed, thermal overload protected industrial motor.
  • Industrial triplex plunger pump with ceramic plungers and stainless steel valves.
  • Low pressure chemical injection.
  • System protected by unloader valve and secondary pressure pop off.
  • Burner system includes a rust-free fuel tank and schedule 80 coil with stainless steel wrap.
  • Adjustable thermostat and temperature limit switch for max 195° F.
  • Heavy duty trigger gun with dual wand and side handle for high pressure and detergent application.
  • Quick change 0°, 15°, 25°, and 40° nozzles.
  • Accessory reel can be added to the machine frame.
Catalog No. 50-283-8308 Supplier MSC Supplier No. 54497367
Add to Cart
Add to Cart
Request a Quote
This portable hot water pressure washer offers economical solutions for various applications. It features a frame with a hose and wand holder and rolls easily on two 13 in wheels.
  • Totally enclosed, thermal overload protected industrial motor.
  • Industrial triplex plunger pump with ceramic plungers and stainless steel valves.
  • Low pressure chemical injection.
  • System protected by unloader valve and secondary pressure pop off.
  • Burner system includes a rust-free fuel tank and schedule 80 coil with stainless steel wrap.
  • Adjustable thermostat and temperature limit switch for max 195° F.
  • Heavy duty trigger gun with dual wand and side handle for high pressure and detergent application.
  • Quick change 0°, 15°, 25°, and 40° nozzles.
  • Accessory reel can be added to the machine frame.