Learn More
Streck Diesse MINI-CUBE ESR Printer
GSA_VASDP
Catalog No. 23285257 Shop All Streck Laboratories Products
Change view
Click to view available options
For Use With (Equipment):
MINI-CUBE ESR Analyzer
Catalog No. For Use With (Equipment)
23-285-257 MINI-CUBE ESR Analyzer
1 options
Catalog No. 23-285-257 Supplier Streck Supplier No. 240402
Add to Cart
Add to Cart
A Diesse MINI-CUBE accessory that connects to the system via bluetooth. Patient and QC results are printed immediately following the conclusion of the test.
  • Bluetooth connection to Diesse MINI-CUBE.
  • Automatically prints Patient and QC results at the conclusion of the test.
Warranty and Services
12 Month Standard Warranty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Printer
For Use With (Application) Automatically prints patient and QC results upon completion of ESR test
For Use With (Equipment) MINI-CUBE ESR Analyzer