Learn More
Streck Diesse MINI-CUBE ESR Scanner
GSA_VASDP
Catalog No. 23285260 Shop All Streck Laboratories Products
Change view
Click to view available options
For Use With (Equipment):
MINI-CUBE ESR Analyzer
Catalog No. For Use With (Equipment)
23-285-260 MINI-CUBE ESR Analyzer
1 options
Catalog No. 23-285-260 Supplier Streck Supplier No. 240405
Add to Cart
Add to Cart
A Diesse MINI-CUBE accessory used to scan patient and QC barcodes to maintain positive sample identification.
  • Ensures positive patient ID.
  • Provides easy QC lot registration and identification of daily QC runs.
Warranty and Services
12 Month Standard Warranty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Barcode Scanner
For Use With (Application) Scans and automatically stores patient identification with ESR result
For Use With (Equipment) MINI-CUBE ESR Analyzer