Learn More
Ankom Technology Dietary Fiber IDF Bag, 50 bags per pack, for Insoluble Dietary Fiber analysis, compatible with ANKOM Dietary Fiber Analyzer, durable filtration material, captures IDF after enzymatic digestion
SDP
Catalog No. NC2209134
Encompass
Dietary Fiber IDF Bag, 50 bags per pack, for Insoluble Dietary Fiber analysis, compatible with ANKOM Dietary Fiber Analyzer, durable filtration material, captures IDF after enzymatic digestion
This item is not returnable. View return policy
Catalog No. NC2209134 Supplier Ankom Technology Supplier No. DFI
Dietary Fiber IDF Bag, 50 bags per pack, for Insoluble Dietary Fiber analysis, compatible with ANKOM Dietary Fiber Analyzer, durable filtration material, captures IDF after enzymatic digestion