Learn More
Dietary Fiber, Total Assay Control Kit, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
- Kit provides standards for use as internal controls in conjunction with the Total Dietary Fiber Assay Kit
- It is suitable for applications including cannabis testing, food and beverage analysis, and general analytical testing.
- Kit Components: Arabinogalactan 1 g
- Tested Techniques: Photometry
- Quantity: Sufficient for ≥10 assays
- Storage Temperature: 2 °C to 8 °C
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| Product Type | Assay Control Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.