Learn More
Abcam 7-Diethylaminocoumarin-3-carbo x ylic acid, amine and protein derivatisation reagent, 100MG
Catalog No. 50996175
Encompass_Preferred
MW 261.27 g/mol. Derivatisation reagent. Fluorescent label for amine modification and protein conjugation. Also used for labeling synthetic peptides for high-throughput detection.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-175 Supplier Abcam Supplier No. AB145332100MG
MW 261.27 g/mol. Derivatisation reagent. Fluorescent label for amine modification and protein conjugation. Also used for labeling synthetic peptides for high-throughput detection.
The product is subject to the following: Abcam Restricted Use Statement